Learn More
Alkali Scientific Pulse 650 Ultrasonic Homogeniz
SDP
Catalog No. 503029651
Encompass_Preferred
Ideal for cell disruption homogenization emulsification and more Large touch screen with total control over operating parameters Supplied complete with controller 6mm horn temperature probe and soundproof enclosure UV-C sterilization and lighted interior of soundproof box Like the smaller Pulse 150 the Pulse 650 utilizes ultrasonic waves and cavitation to disrupt cells shear DNA and create emulsions With up to 650 watts of power samples from 0 1ml to 500ml can be processed Pulsed operation power ratios and other operating parameters are easily set using the large touch screen Alarms alert the user to over temperature over/no load and expiration of time The Pulse 650 is complete with everything needed for operation including a soundproof enclosure for reducing noise levels The low-profile controller and soundproof enclosure are stackable to save valuable bench space The enclosure features interior lights and a large front panel for viewing samples A remote controlled motorized l
Catalog No. 50-302-9651 Supplier Alkali Scientific Supplier No. RS0650
Ideal for cell disruption homogenization emulsification and more Large touch screen with total control over operating parameters Supplied complete with controller 6mm horn temperature probe and soundproof enclosure UV-C sterilization and lighted interior of soundproof box Like the smaller Pulse 150 the Pulse 650 utilizes ultrasonic waves and cavitation to disrupt cells shear DNA and create emulsions With up to 650 watts of power samples from 0 1ml to 500ml can be processed Pulsed operation power ratios and other operating parameters are easily set using the large touch screen Alarms alert the user to over temperature over/no load and expiration of time The Pulse 650 is complete with everything needed for operation including a soundproof enclosure for reducing noise levels The low-profile controller and soundproof enclosure are stackable to save valuable bench space The enclosure features interior lights and a large front panel for viewing samples A remote controlled motorized l