Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Pulse Dampener for L/S and I/P Tubing
Catalog No. NC9429941
Encompass
This pulse damper virtually eliminates pulsation in output flow. Featuring a polyethylene body with five pairs of fittings and a PTFE-pipe thread tape are included. Fittings included are (tubing ID x NPT(M) thread); 1/16" x 1/8", 1/8" x 1/8", 3/16" x 1/8", 1/4" x 1/8", and 3/8" x 1/8".
Catalog No. NC9429941 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0759620
This pulse damper virtually eliminates pulsation in output flow. Featuring a polyethylene body with five pairs of fittings and a PTFE-pipe thread tape are included. Fittings included are (tubing ID x NPT(M) thread); 1/16" x 1/8", 1/8" x 1/8", 3/16" x 1/8", 1/4" x 1/8", and 3/8" x 1/8".