Learn More
Waters Corp Waters Corporation Pulse Dampener Rebuild Kit, For 1500 Series HPLC Pumps, Repairs Leaks
Catalog No. 50792321 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Pulse Dampener Rebuild Kit is categorized under Service Parts & Kits, signifying its role in the maintenance and repair of instrumentation. It is specifically designed to address suspected blockages or leaks in 1500 Series HPLC Pumps, thereby restoring their optimal operation.
  • Purpose-built for 1500 series HPLC pumps.
  • Designed to address suspected blockages or leaks.
  • Restores proper pump operation.
  • Facilitates testing procedures to identify leaks or plugs.
  • Provides components for the transducer, draw-off valve, and mixing tee connections.
  • Supports back-flushing capability with polar solvents.
Catalog No. 50-792-321 Supplier Waters Corp Supplier No. 700000163
This Pulse Dampener Rebuild Kit is categorized under Service Parts & Kits, signifying its role in the maintenance and repair of instrumentation. It is specifically designed to address suspected blockages or leaks in 1500 Series HPLC Pumps, thereby restoring their optimal operation.
  • Purpose-built for 1500 series HPLC pumps.
  • Designed to address suspected blockages or leaks.
  • Restores proper pump operation.
  • Facilitates testing procedures to identify leaks or plugs.
  • Provides components for the transducer, draw-off valve, and mixing tee connections.
  • Supports back-flushing capability with polar solvents.