Learn More
MSC Guardair Dust Extractor Kit, 30 Gallon, 1.5 in Inlet, Compressed Air
Catalog No. 502838111
Encompass
This PulseAir Dust Extractor Kit is engineered for exceptional performance in pneumatic vacuum technology, ideal for use with source-capture tools. It features button-activated filter cleaning and advanced noise reduction, operating quietly at 88 dBA or less. Powered by compressed air, this unit minimizes operator exposure to harmful dust and meets OSHA Silica Dust Regulation.
  • Consumes up to 70% less compressed air for high vacuum suction
  • Customizable performance with three injector nozzle sets
  • Proprietary diecast aluminum, 2-stage, multi-venturi vacuum-generating engine
  • Backwards compatible with existing pneumatic vacuum components
  • Electrically bonded components eliminate static electricity and shocks
  • Equipped with a liquid shutoff assembly for wet and dry applications
Catalog No. 50-283-8111 Supplier MSC Supplier No. 64358773
This PulseAir Dust Extractor Kit is engineered for exceptional performance in pneumatic vacuum technology, ideal for use with source-capture tools. It features button-activated filter cleaning and advanced noise reduction, operating quietly at 88 dBA or less. Powered by compressed air, this unit minimizes operator exposure to harmful dust and meets OSHA Silica Dust Regulation.
  • Consumes up to 70% less compressed air for high vacuum suction
  • Customizable performance with three injector nozzle sets
  • Proprietary diecast aluminum, 2-stage, multi-venturi vacuum-generating engine
  • Backwards compatible with existing pneumatic vacuum components
  • Electrically bonded components eliminate static electricity and shocks
  • Equipped with a liquid shutoff assembly for wet and dry applications