Learn More
MSC Guardair Wet/Dry Vacuum, Air, 20 gal, HEPA, Steel Tank
Catalog No. 502838024
Encompass
This air-powered wet/dry vacuum offers button-activated filter cleaning and electrically bonded components for static prevention. It minimizes dust exposure, meeting OSHA Silica Dust Regulation, and operates quietly without electrical parts, providing significant cost savings.
  • Button-activated, on-demand filter cleaning for ease of use.
  • Minimizes operator dust exposure and meets OSHA Silica Dust Regulation.
  • Electrically bonded components and static conductive attachments prevent static and shocks.
  • Compressed air powered, with no rotating parts, motors to spark, or electric cords.
  • Consumes up to 70% less compressed air while providing superior vacuum suction.
  • Ultra-quiet operation at 88 dBA or less.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
Catalog No. 50-283-8024 Supplier MSC Supplier No. 64357353
This air-powered wet/dry vacuum offers button-activated filter cleaning and electrically bonded components for static prevention. It minimizes dust exposure, meeting OSHA Silica Dust Regulation, and operates quietly without electrical parts, providing significant cost savings.
  • Button-activated, on-demand filter cleaning for ease of use.
  • Minimizes operator dust exposure and meets OSHA Silica Dust Regulation.
  • Electrically bonded components and static conductive attachments prevent static and shocks.
  • Compressed air powered, with no rotating parts, motors to spark, or electric cords.
  • Consumes up to 70% less compressed air while providing superior vacuum suction.
  • Ultra-quiet operation at 88 dBA or less.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.