Learn More
MSC Guardair Dust Extractor Kit, 30 Gallon, 2 inch Inlet, PulseAir, Pneumatic
Catalog No. 502838094
Encompass
This PowerQUAD 30-Gallon PulseAir Dust Extractor Kit is a pneumatic vacuum system for high performance with source-capture tools. It offers on-demand filter cleaning, minimizing operator exposure to harmful dust and meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Powered by compressed air, it eliminates motors and electric cords, providing superior vacuum suction and significant operating cost savings.
  • Consumes less compressed air for high vacuum suction and operating cost savings.
  • Operates quietly at 88 dBA or less, 10-15 dB quieter than traditional vacuums.
  • Three color-coded injector nozzle sets allow tailored performance.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Compatible with existing pneumatic vacuum components.
  • Electrically bonded components and grounding eliminate static electricity.
Catalog No. 50-283-8094 Supplier MSC Supplier No. 64359011
This PowerQUAD 30-Gallon PulseAir Dust Extractor Kit is a pneumatic vacuum system for high performance with source-capture tools. It offers on-demand filter cleaning, minimizing operator exposure to harmful dust and meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Powered by compressed air, it eliminates motors and electric cords, providing superior vacuum suction and significant operating cost savings.
  • Consumes less compressed air for high vacuum suction and operating cost savings.
  • Operates quietly at 88 dBA or less, 10-15 dB quieter than traditional vacuums.
  • Three color-coded injector nozzle sets allow tailored performance.
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum engine.
  • Compatible with existing pneumatic vacuum components.
  • Electrically bonded components and grounding eliminate static electricity.