Learn More
MSC Guardair Dust Extractor Kit, 20 Gallon, PulseAir, 2 inch Inlet
Catalog No. 502837019
Encompass
This pneumatic dust extractor kit offers exceptional performance for source-capture tools, with button-activated filter cleaning and minimal operator exposure to harmful dust, meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Electrically bonded components and static conductive attachments eliminate static electricity. Powered by compressed air, it has no rotating parts or electric cords, generating superior vacuum suction and operating cost savings.
  • Energy efficient and powerful, with high vacuum suction and operating cost savings.
  • Advanced noise reduction, operating quietly at 88 dBA or less.
  • Customizable performance via three color-coded injector nozzle sets.
  • Proprietary diecast aluminum, multi-venturi vacuum generating engine.
  • Compatible with existing pneumatic vacuum components.
Catalog No. 50-283-7019 Supplier MSC Supplier No. 64357643
This pneumatic dust extractor kit offers exceptional performance for source-capture tools, with button-activated filter cleaning and minimal operator exposure to harmful dust, meeting OSHA Silica Dust Regulation 29 CFR 1926.1153. Electrically bonded components and static conductive attachments eliminate static electricity. Powered by compressed air, it has no rotating parts or electric cords, generating superior vacuum suction and operating cost savings.
  • Energy efficient and powerful, with high vacuum suction and operating cost savings.
  • Advanced noise reduction, operating quietly at 88 dBA or less.
  • Customizable performance via three color-coded injector nozzle sets.
  • Proprietary diecast aluminum, multi-venturi vacuum generating engine.
  • Compatible with existing pneumatic vacuum components.