Learn More
Fritsch Milling And Sizing Inc PULVERISETTE 19 CUTTING MILL
SDP
Catalog No. NC3706292
Encompass_Preferred
  • Rotational speed of 50-700 rpm, variable adjustable
  • Torque of 26 Nm
  • Power of 2350 watt
  • Voltage of 200-240 V/1~
  • Frequency of 50-60 Hz
  • Cutting working principle
  • Targeting materials: Hard, tough-elastic, fibrous, temperature-sensitive materials, and plastics
  • Final fineness range of 0.1–20 mm (dependent on sieve cassette)
  • Optimized clean design for ease of cleaning
  • Maintenance-free three-phase motor with frequency converter
  • Grinding chamber with minimal dead space and progressive cutting geometry
  • Drive stability enhanced by flywheel mass
  • Optional corrosion resistance with stainless steel 316L version available
Catalog No. NC3706292 Supplier Fritsch Milling And Sizing Inc Supplier No. 19.3000.00
Add to Cart
Add to Cart
Request a Quote
  • Rotational speed of 50-700 rpm, variable adjustable
  • Torque of 26 Nm
  • Power of 2350 watt
  • Voltage of 200-240 V/1~
  • Frequency of 50-60 Hz
  • Cutting working principle
  • Targeting materials: Hard, tough-elastic, fibrous, temperature-sensitive materials, and plastics
  • Final fineness range of 0.1–20 mm (dependent on sieve cassette)
  • Optimized clean design for ease of cleaning
  • Maintenance-free three-phase motor with frequency converter
  • Grinding chamber with minimal dead space and progressive cutting geometry
  • Drive stability enhanced by flywheel mass
  • Optional corrosion resistance with stainless steel 316L version available