Learn More
Aviva Systems Biology Corp PUM2 Antibody - N-terminal region (ARP40810_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40810_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PUM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PUM2. Alternate names: KIAA0235, PUMH2, PUML2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57629