Learn More
Aviva Systems Biology Corp PUM2 Antibody - N-terminal region (ARP40811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PUM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PUM2. Alternate names: FLJ36528, KIAA0235, MGC138251, MGC138253, PUMH2, PUML2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57630