Learn More
Cayman Chemical PUMA BH3humntrIfluoroac 10mg
SDP
Catalog No. 502915623
Encompass_Preferred
A peptide activator of Bak; binds to Bak (Kds = 290 and 26 nM in HEPES and CHAPS buffer, respectively) and induces Bak homo-oligomerization and Bak-mediated membrane permeabilization
Catalog No. 50-291-5623 Supplier Cayman Chemical Supplier No. 3750910 MG
A peptide activator of Bak; binds to Bak (Kds = 290 and 26 nM in HEPES and CHAPS buffer, respectively) and induces Bak homo-oligomerization and Bak-mediated membrane permeabilization