Learn More
Electron Microscopy Sciences Flexible Shaft Machine Pumice 72 Wheel Assortment
SDP
Catalog No. 50996828
Encompass_Preferred
For cleaning, polishing and finishing. Made from natural rubber with graded pumice grain distributed to assure uniform results. Kit includes 12 discs each of six styles and 3 mandrels with 1/8" (3.18mm) shanks.
Catalog No. 50-996-828 Supplier Electron Microscopy Sciences Supplier No. 6214372
For cleaning, polishing and finishing. Made from natural rubber with graded pumice grain distributed to assure uniform results. Kit includes 12 discs each of six styles and 3 mandrels with 1/8" (3.18mm) shanks.