Learn More
MSC Permatex Hand Cleaner, Fine Pumice, 1 Gallon, Pump Bottle
Catalog No. 502837106
Encompass
This biodegradable hand cleaner features MicroGel™ technology for effective cleaning without petroleum solvents. It has a fresh citrus scent and is fortified with skin conditioners like aloe vera, lanolin, propylene glycol, jojoba oil, and vitamin E. The fine pumice lotion formula is designed for deep cleaning, while leaving hands smooth and not sticky.
  • Removes grease, grime, oils, soil, gasket cement, glues, paints, and inks.
  • Cleans hands without water.
  • Contains biodegradable surfactants.
  • Leaves hands smooth, not sticky.
  • Ultra low VOC.
  • Improved environmental, health, and safety profile.
  • Contains extra fine pumice for deep cleaning.
  • Pleasant orange scent.
Catalog No. 50-283-7106 Supplier MSC Supplier No. 63429419
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This biodegradable hand cleaner features MicroGel™ technology for effective cleaning without petroleum solvents. It has a fresh citrus scent and is fortified with skin conditioners like aloe vera, lanolin, propylene glycol, jojoba oil, and vitamin E. The fine pumice lotion formula is designed for deep cleaning, while leaving hands smooth and not sticky.
  • Removes grease, grime, oils, soil, gasket cement, glues, paints, and inks.
  • Cleans hands without water.
  • Contains biodegradable surfactants.
  • Leaves hands smooth, not sticky.
  • Ultra low VOC.
  • Improved environmental, health, and safety profile.
  • Contains extra fine pumice for deep cleaning.
  • Pleasant orange scent.