Learn More
Medchemexpress LLC Pumosetrag Hydrochloride | 194093-42-0 | 99.8% | 339.84 | 1 ML
SDP
Catalog No. 5000401302
Encompass_Preferred
Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Orally available 5-HT3 partial agonist
  • Developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease
  • Activity profile varies by species and region of the intestine
  • Inhibits responses to ondansetron, confirming action on 5-HT3 receptors
  • Delays liquid gastric emptying, stimulates fasting antroduodenal migrating motor complex activity, and accelerates small intestinal transit
Catalog No. 50-004-01302 Supplier Medchemexpress LLC Supplier No. HY1965010MM/1ML
Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Orally available 5-HT3 partial agonist
  • Developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease
  • Activity profile varies by species and region of the intestine
  • Inhibits responses to ondansetron, confirming action on 5-HT3 receptors
  • Delays liquid gastric emptying, stimulates fasting antroduodenal migrating motor complex activity, and accelerates small intestinal transit