Learn More
Medchemexpress LLC Pumosetrag Hydrochloride | 194093-42-0 | 99.8% | 339.84 | 25 MG
SDP
Catalog No. 5000402663
Encompass_Preferred
Pumosetrag Hydrochloride is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease. It is intended for research use only.
  • Orally available 5-HT3 partial agonist
  • Developed for treatment of irritable bowel syndrome and gastroesophageal reflux disease
  • Exhibits regional and species specificities
  • Delays liquid gastric emptying
  • Relaxes the proximal stomach
  • Stimulates fasting antroduodenal migrating motor complex activity
  • Accelerates small intestinal transit
Catalog No. 50-004-02663 Supplier Medchemexpress LLC Supplier No. 25MGHY19650
Pumosetrag Hydrochloride is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease. It is intended for research use only.
  • Orally available 5-HT3 partial agonist
  • Developed for treatment of irritable bowel syndrome and gastroesophageal reflux disease
  • Exhibits regional and species specificities
  • Delays liquid gastric emptying
  • Relaxes the proximal stomach
  • Stimulates fasting antroduodenal migrating motor complex activity
  • Accelerates small intestinal transit