Learn More
Medchemexpress LLC Pumosetrag Hydrochloride | 194093-42-0 | 99.8% | 339.84 | 50 MG
SDP
Catalog No. 5000400499
Encompass_Preferred
Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Orally available 5-HT3 partial agonist.
  • Developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Displays regional and species specificities.
  • Activity is inhibited by ondansetron, confirming action on 5-HT(3) receptors.
  • Delays liquid gastric emptying, stimulates fasting antroduodenal migrating motor complex activity, and accelerates small intestinal transit.
Catalog No. 50-004-00499 Supplier Medchemexpress LLC Supplier No. HY1965050MG
Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Orally available 5-HT3 partial agonist.
  • Developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease.
  • Displays regional and species specificities.
  • Activity is inhibited by ondansetron, confirming action on 5-HT(3) receptors.
  • Delays liquid gastric emptying, stimulates fasting antroduodenal migrating motor complex activity, and accelerates small intestinal transit.