Learn More
Medchemexpress LLC Pumosetrag hydrochloride | 194093-42-0 | MFCD00939557 | 99.8% | 339.84 g/mol | C15H18ClN3O2S | 10 MG
SDP
Catalog No. 5000294829
Encompass_Preferred
Pumosetrag hydrochloride is an orally active, small-molecule partial agonist of the 5-HT3 receptor investigated for treatment of irritable bowel syndrome and gastroesophageal reflux disease. Supplied as a high-purity white to off-white solid for laboratory research (for research use only).
  • Acts as a partial agonist at 5-HT3 receptors.
  • Investigated for irritable bowel syndrome and gastroesophageal reflux disease.
  • Supplied as a white to off-white solid with high purity.
  • Molecular weight 339.84 g/mol; formula C15H18ClN3O2S.
  • Available in small laboratory pack sizes, including 10 mg.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-94829 Supplier Medchemexpress LLC Supplier No. HY1965010MG
Pumosetrag hydrochloride is an orally active, small-molecule partial agonist of the 5-HT3 receptor investigated for treatment of irritable bowel syndrome and gastroesophageal reflux disease. Supplied as a high-purity white to off-white solid for laboratory research (for research use only).
  • Acts as a partial agonist at 5-HT3 receptors.
  • Investigated for irritable bowel syndrome and gastroesophageal reflux disease.
  • Supplied as a white to off-white solid with high purity.
  • Molecular weight 339.84 g/mol; formula C15H18ClN3O2S.
  • Available in small laboratory pack sizes, including 10 mg.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.