Learn More
Medchemexpress LLC HY-19650 5mg Medchemexpress, Pumosetrag Hydrochloride CAS:194093-42-0 Purity:>98%
SDP
Catalog No. 502082159
Encompass_Preferred
Medchemexpress, HY-19650 5mg Pumosetrag Hydrochloride CAS:194093-42-0 Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2159 Supplier Medchemexpress LLC Supplier No. HY196505MG
Medchemexpress, HY-19650 5mg Pumosetrag Hydrochloride CAS:194093-42-0 Pumosetrag Hydrochloride (MKC-733; DDP-733) is an orally available 5-HT3 partial agonist developed for the treatment of irritable bowel syndrome and gastroesophageal reflux disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.