Learn More
Instech Syringe pump, Harvard Apparatus model 11 Pico Plus Elite infuse/withdraw programmable single syringe pump (704511), US cord
SDP
Catalog No. NC2816861
Encompass
The Pump 11 Elite expands the capabilities of the original workhorse Pump 11 series with a high resolution touch screen and intuitive icon interface. The interface allows you to create, save and run simple to complex methods without a PC.
The Pump 11 Elite include syringe clamp for syringes ranging from 0.5µl to 60ml (single syringe) and 0.5µl to 10 ml (dual syringe). Offers accuracy from 0.58 picoliters/min up to 88.28 ml/min, depending on the model and syringe size.
The Pump 11 Elite is available in Infusion Only or Infusion/Withdrawal Programmable Models with single or dual syringe racks. These pumps have advanced connectivity with a USB port for computer control, a footswitch, and, on programmable models, RS-485 ports for pump-to-pump connections and digital I/O for remote control.
Note that these pumps do not have an occlusion alarm or pressure monitor, which are recommended when infusing laboratory animals for animal welfare and data quality.
Catalog No. NC2816861 Supplier Instech Supplier No. HA1100U
The Pump 11 Elite expands the capabilities of the original workhorse Pump 11 series with a high resolution touch screen and intuitive icon interface. The interface allows you to create, save and run simple to complex methods without a PC.
The Pump 11 Elite include syringe clamp for syringes ranging from 0.5µl to 60ml (single syringe) and 0.5µl to 10 ml (dual syringe). Offers accuracy from 0.58 picoliters/min up to 88.28 ml/min, depending on the model and syringe size.
The Pump 11 Elite is available in Infusion Only or Infusion/Withdrawal Programmable Models with single or dual syringe racks. These pumps have advanced connectivity with a USB port for computer control, a footswitch, and, on programmable models, RS-485 ports for pump-to-pump connections and digital I/O for remote control.
Note that these pumps do not have an occlusion alarm or pressure monitor, which are recommended when infusing laboratory animals for animal welfare and data quality.