Learn More
Icon Scientific Inc PUMP 80P W/1000 ML PUMP HEAD
SDP
Catalog No. NC3106199
Encompass
  • High-performance preparative pump for HPLC applications.
  • Compatible with a wide range of solvents.
  • Flow rate range of 1-1000 ml/min.
  • Best working conditions at 2-800 ml/min.
  • Continuous working conditions of 2-400 ml/min.
  • Uses active pulsation compensation for stable flow.
  • Features active piston seal washing for enhanced longevity.
  • Modular design allows for various gradient configurations.
  • Integrated RFID for automatic pump head recognition.
  • Liquid temperature range from 4-60 °C.
  • Dimensions: 361 mm H x 208.2 mm W x 523 mm D.
  • Weight: 19 kg.
  • Power supply: 100-240 V, 50-60 Hz.
Catalog No. NC3106199 Supplier Icon Scientific Inc Supplier No. APD30NA
Add to Cart
Add to Cart
Request a Quote
  • High-performance preparative pump for HPLC applications.
  • Compatible with a wide range of solvents.
  • Flow rate range of 1-1000 ml/min.
  • Best working conditions at 2-800 ml/min.
  • Continuous working conditions of 2-400 ml/min.
  • Uses active pulsation compensation for stable flow.
  • Features active piston seal washing for enhanced longevity.
  • Modular design allows for various gradient configurations.
  • Integrated RFID for automatic pump head recognition.
  • Liquid temperature range from 4-60 °C.
  • Dimensions: 361 mm H x 208.2 mm W x 523 mm D.
  • Weight: 19 kg.
  • Power supply: 100-240 V, 50-60 Hz.