Learn More
Ace Glass, Inc. Pump, vacuum, chemical resistant diaphragm, 3 stage, 1.5 torr ultimate vacuum, 37L/min flow rate.
SDP
Catalog No. 50978145
Encompass
Pump, vacuum, chemical resistant diaphragm, 3 stage, 1.5 torr ultimate vacuum, 37L/min flow rate.
Catalog No. 50-978-145 Supplier Ace Glass, Inc. Supplier No. 1411215
Add to Cart
Add to Cart
Request a Quote
Pump, vacuum, chemical resistant diaphragm, 3 stage, 1.5 torr ultimate vacuum, 37L/min flow rate.