Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Analog, Variable-Speed, Modular Drive, 6 to 600 rpm, 115 VAC
Catalog No. NC1092634
Encompass
A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range, ±2% speed regulation, and ETL and CE listings. For added versatility, drives accept all L/S pump heads.
Catalog No. NC1092634 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0755900
A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range, ±2% speed regulation, and ETL and CE listings. For added versatility, drives accept all L/S pump heads.