Learn More
Waters Corp Pump Head 2545, 0.50 to 150.00 mL/min, 6000 psi, 316 Stainless Steel
Catalog No. 50792592 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pump head is a core component of the 2545 Quaternary Gradient Module, which functions as a solvent delivery device for purification systems, operating as a low-pressure mixing quaternary gradient pump.
  • Flow-rate range: 0.50 to 150.00 mL/min, in 0.01 mL/min increments
  • Maximum operating pressure: 6000 psi up to 100 mL/min; rolls off to 5000 psi at 150 mL/min
  • Automatic and continuous compressibility compensation
  • Integral, active, programmable plunger seal wash
  • Primary wetted materials include 316 stainless steel, UHMWPE blend, sapphire, ruby, Tefzel (ETFE), Teflon (FEP, PTFE, PFA), Valcon H, Nitronic 60, and PEEK
  • Flow accuracy: ± 1.0% of required flow (methanol or water at 600 psi)
Catalog No. 50-792-592 Supplier Waters Corp Supplier No. 700001526
This pump head is a core component of the 2545 Quaternary Gradient Module, which functions as a solvent delivery device for purification systems, operating as a low-pressure mixing quaternary gradient pump.
  • Flow-rate range: 0.50 to 150.00 mL/min, in 0.01 mL/min increments
  • Maximum operating pressure: 6000 psi up to 100 mL/min; rolls off to 5000 psi at 150 mL/min
  • Automatic and continuous compressibility compensation
  • Integral, active, programmable plunger seal wash
  • Primary wetted materials include 316 stainless steel, UHMWPE blend, sapphire, ruby, Tefzel (ETFE), Teflon (FEP, PTFE, PFA), Valcon H, Nitronic 60, and PEEK
  • Flow accuracy: ± 1.0% of required flow (methanol or water at 600 psi)