Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load II Pump Head for High-Performance Precision Tubing, PPS Housing, CRS Rotor
Catalog No. NC9328987
Encompass
These pump heads accept the four L/S high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
Catalog No. NC9328987 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720052
These pump heads accept the four L/S high-performance precision tubing sizes. High-performance precision tubing has a thicker wall than precision tubing enabling better pressure generation, greater suction lift, and greater ability to pump viscous fluids. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.