Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Multichannel Cartridge Pump Head for Microbore and Precision Tubing, 8-channels, 4-rollers
Catalog No. NC9684834
Encompass
Each pump head system is composed of a heavy-duty pump head and two sizes of snap-in tubing cartridges, each with individually adjustable occlusion settings (order cartridges separately). The occlusion "wedge" design provides accurate and more balanced flow control. Recommended for use with small cartridge 07519-80 and large cartridge 07519-70.
Catalog No. NC9684834 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0751906
Each pump head system is composed of a heavy-duty pump head and two sizes of snap-in tubing cartridges, each with individually adjustable occlusion settings (order cartridges separately). The occlusion "wedge" design provides accurate and more balanced flow control. Recommended for use with small cartridge 07519-80 and large cartridge 07519-70.