Learn More
Waters Corp Pump Head, Titanium, ACQUITY H-Class Bio, for BioQSM
Catalog No. 502392046 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pump head is a crucial component for UPLC systems, specifically designed for the ACQUITY H-Class Bio and ACQUITY Arc Premier systems. It is constructed from titanium, making it suitable for biocompatible applications and providing enhanced chemical resistance.
  • Constructed from titanium for biocompatible applications and enhanced chemical resistance.
  • Compatible with BSM and QSM components.
  • Designed for H-Class and Arc Premier system instruments.
  • Supplied as one unit per package.
  • Replacement is on an as-required basis.
Catalog No. 50-239-2046 Supplier Waters Corp Supplier No. 700005411
This pump head is a crucial component for UPLC systems, specifically designed for the ACQUITY H-Class Bio and ACQUITY Arc Premier systems. It is constructed from titanium, making it suitable for biocompatible applications and providing enhanced chemical resistance.
  • Constructed from titanium for biocompatible applications and enhanced chemical resistance.
  • Compatible with BSM and QSM components.
  • Designed for H-Class and Arc Premier system instruments.
  • Supplied as one unit per package.
  • Replacement is on an as-required basis.