Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Standard Pump Head for Precision Tubing L/S 13, Polycarbonate Housing, CRS Rotor
Catalog No. NC1807543
Encompass
This Standard pump head accepts precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.
Catalog No. NC1807543 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701320
This Standard pump head accepts precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.