Learn More
Waters Corp Waters Pump Head Plunger Seal, .0787 ID, Flanged, UP30 Seal, 2 pk
Catalog No. 502039440 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement high pressure plunger seal. It is found in pump heads on ACQUITY Quaternary Solvent Managers and ACQUITY UPLC Binary and Isocratic and NanoACQUITY Solvent Managers. It is also found in the UPC2 Convergence Manager.
  • Designed as a replacement high pressure plunger seal.
  • Features a .0787 ID, flanged design, and UP30 seal.
  • Provided with 2 units per package.
  • Compatible components include ACQUITY UPLC ISM, ACQUITY UPLC BSM, ACQUITY UPLC QSM, ACQUITY UPLC H-Class QSM, and nanoACQUITY BSM.
  • Suitable for UPLC system types.
  • Can be used with ACQUITY UPLC H-Class System, nanoACQUITY UPLC System, ACQUITY, ACQUITY Premier, and UPC2 instruments.
  • Recommended for annual replacement.
Catalog No. 50-203-9440 Supplier Waters Corp Supplier No. 700002599
This is a replacement high pressure plunger seal. It is found in pump heads on ACQUITY Quaternary Solvent Managers and ACQUITY UPLC Binary and Isocratic and NanoACQUITY Solvent Managers. It is also found in the UPC2 Convergence Manager.
  • Designed as a replacement high pressure plunger seal.
  • Features a .0787 ID, flanged design, and UP30 seal.
  • Provided with 2 units per package.
  • Compatible components include ACQUITY UPLC ISM, ACQUITY UPLC BSM, ACQUITY UPLC QSM, ACQUITY UPLC H-Class QSM, and nanoACQUITY BSM.
  • Suitable for UPLC system types.
  • Can be used with ACQUITY UPLC H-Class System, nanoACQUITY UPLC System, ACQUITY, ACQUITY Premier, and UPC2 instruments.
  • Recommended for annual replacement.