Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Easy-Load Pump Head for Precision Tubing, Black PPS Housing, SS Rotor
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7760160
Encompass
The Masterflex I/P Easy-Load pump head combines an over-center cam with automatic tubing retention in a rugged, chemical-resistant housing. This design allows fast, easy tubing changes without removing the pump head from the drive. Built-in tubing guide ensures correct positioning of tubing to maximize performance and pump tubing life. Spring-loaded tubing retainers ensure tubing is secured automatically to promote long tubing life and optimize performance of each tubing size. Select from Polyphenylene Sulfide (PPS) housing with CRS or SS rotor. SS rotors feature PTFE-sealed bearings for long life and reduced maintenance; CRS rotors have Buna N shielded bearings. Dual-channel hardware allows two pump heads to operate on a single drive¿order hardware set 77601-96 separately. With single or multiple channels, tubing can be changed in each channel without removing the pump heads from your drive.
Catalog No. NC9666855