Learn More
BURKLE INC Pump-it canister pump, PP/FKM, pump cap. 2.11 gal. (8l)/min
Catalog No. 5000025841
Encompass_Preferred
The Pump-it canister pump is ideal for transferring small liquid amounts from canisters up to 60 liters (15.85 gal). Safe and quick filling and emptying of canisters, carboys, and small drums.
Its acid-proof design seals containers airtight, allowing the pump to stay attached and ready for use anytime.
Repeated hand piston action builds pressure, and opening the stopcock releases fluid with a smooth, pulsation-free flow for precise dosing. Closing stops flow immediately without run-on.
The adjustable delivery tube extends from 550 to 39.37 in. (1000 mm) (21.65 to 39.37 in) for full container emptying and can be shortened for smaller containers.
Lightweight and compact, suitable for weak acids, alkalis, and petrochemicals.
Catalog No. 50-000-25841 Supplier BURKLE INC Supplier No. 52011000
The Pump-it canister pump is ideal for transferring small liquid amounts from canisters up to 60 liters (15.85 gal). Safe and quick filling and emptying of canisters, carboys, and small drums.
Its acid-proof design seals containers airtight, allowing the pump to stay attached and ready for use anytime.
Repeated hand piston action builds pressure, and opening the stopcock releases fluid with a smooth, pulsation-free flow for precise dosing. Closing stops flow immediately without run-on.
The adjustable delivery tube extends from 550 to 39.37 in. (1000 mm) (21.65 to 39.37 in) for full container emptying and can be shortened for smaller containers.
Lightweight and compact, suitable for weak acids, alkalis, and petrochemicals.