Learn More
AVANTOR MASTERFLEX (VWR LLC) PUMP MFLEX I/P EIP 650R EL
Catalog No. NC3308885
Encompass
  • Robust and versatile peristaltic pump system for precise fluid transfer
  • Compatible with EtherNet/IP and MasterflexLive cloud-enabled platform
  • Intuitive 7-inch capacitive touchscreen interface for easy monitoring
  • 316 stainless steel housing with IP66/NEMA 4X rating ensures washdown capability
  • I/P Easy-Load pump head for quick and tool-free tubing changes
  • Programmable dispensing with anti-drip function for reliability
  • Reversible motor and self-priming capabilities enhance operational safety
  • Flow rate range: 1 to 8000 ml/min (0.001 to 8 L/min)
  • Speed range: 0.1 to 650 rpm
  • Dimensions: 346 mm x 432 mm x 260 mm (13.6 in x 17 in x 10.2 in)
  • Certifications: UL, cUL, CE Compliant
  • 2-year warranty
Catalog No. NC3308885 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7796600
Add to Cart
Add to Cart
Request a Quote
  • Robust and versatile peristaltic pump system for precise fluid transfer
  • Compatible with EtherNet/IP and MasterflexLive cloud-enabled platform
  • Intuitive 7-inch capacitive touchscreen interface for easy monitoring
  • 316 stainless steel housing with IP66/NEMA 4X rating ensures washdown capability
  • I/P Easy-Load pump head for quick and tool-free tubing changes
  • Programmable dispensing with anti-drip function for reliability
  • Reversible motor and self-priming capabilities enhance operational safety
  • Flow rate range: 1 to 8000 ml/min (0.001 to 8 L/min)
  • Speed range: 0.1 to 650 rpm
  • Dimensions: 346 mm x 432 mm x 260 mm (13.6 in x 17 in x 10.2 in)
  • Certifications: UL, cUL, CE Compliant
  • 2-year warranty