Learn More
AVANTOR MASTERFLEX (VWR LLC) PUMP MFLX B/T 321R E/IP CONS
Catalog No. 502742134
Encompass
Masterflex B/T MasterSense™ BioPharma Pump, EtherNet/IP™ and Profibus Network-Compatible, 11 to 321 rpm; 90 to 260 VAC, Avantor
Catalog No. 50-274-2134 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7711210
Add to Cart
Add to Cart
Request a Quote
Masterflex B/T MasterSense™ BioPharma Pump, EtherNet/IP™ and Profibus Network-Compatible, 11 to 321 rpm; 90 to 260 VAC, Avantor