Learn More
AVANTOR MASTERFLEX (VWR LLC) Pump, Digital Miniflex, Single-Channel, 300 RPM, 115/230 VAC
Catalog No. NC1094265
Encompass
Masterflex L/S Digital Miniflex Pump, Single-Channel, 300 rpm, 90 to 260 VAC, A small footprint digital drive that delivers accurate flow over a wide rangeRequires less than half the bench space compared to the L/S standard digital driveIcon-driven menu allows quick, intuitive programming and setupFlow range of 0.005 to 381 mL/min (depending on model and tubing size selected)Open-head sensor stops the drive when the pump head is opened
Catalog No. NC1094265 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0752520
Masterflex L/S Digital Miniflex Pump, Single-Channel, 300 rpm, 90 to 260 VAC, A small footprint digital drive that delivers accurate flow over a wide rangeRequires less than half the bench space compared to the L/S standard digital driveIcon-driven menu allows quick, intuitive programming and setupFlow range of 0.005 to 381 mL/min (depending on model and tubing size selected)Open-head sensor stops the drive when the pump head is opened