Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Digital Drive with Open-Head Sensor and Easy-Load II Pump Head for Precision Tubing, 600 rpm; 115/230 VAC
Catalog No. NC1869934
Encompass
Masterflex L/S Digital Drive with Open-Head Sensor and Easy-Load II Pump Head for Precision Tubing, 600 rpm; 115/230 VAC,INCLUDESL/S drive 07522-28, Easy-Load II pump head 77200-30, 10-ft C-Flex ULTRA tubing, and 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please specify ultimate destination when ordering.KEY FEATURESDigital interface with graphical LCD shows pump performance and operating modes¿continuous run, timed dispense, copy dispense, or volume dispenseSelect your preferred language from the user-friendly menu for programming and operationDispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy¿1 to 9,999 dispense cycles; or Time¿time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures dispensing accuracy
Catalog No. NC1869934 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7792177
Masterflex L/S Digital Drive with Open-Head Sensor and Easy-Load II Pump Head for Precision Tubing, 600 rpm; 115/230 VAC,INCLUDESL/S drive 07522-28, Easy-Load II pump head 77200-30, 10-ft C-Flex ULTRA tubing, and 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please specify ultimate destination when ordering.KEY FEATURESDigital interface with graphical LCD shows pump performance and operating modes¿continuous run, timed dispense, copy dispense, or volume dispenseSelect your preferred language from the user-friendly menu for programming and operationDispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy¿1 to 9,999 dispense cycles; or Time¿time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures dispensing accuracy