Learn More
AVANTOR MASTERFLEX (VWR LLC) PUMP MFLX LS SS 250 ULTRATHN
Catalog No. 502742117
Encompass
Masterflex UltraPharm™ Filling / Dispensing Pump for L/S Precision Double-Y Tube Sets; 90 to 260 VAC, Avantor
Catalog No. 50-274-2117 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7797640
Add to Cart
Add to Cart
Request a Quote
Masterflex UltraPharm™ Filling / Dispensing Pump for L/S Precision Double-Y Tube Sets; 90 to 260 VAC, Avantor