Learn More
Interscience Pump Module
SDP
Catalog No. 16948961 Shop All Interscience Products
Change view
Click to view available options
For Use With (Equipment):
DiluFlow Elite
Catalog No. For Use With (Equipment)
16-948-961 DiluFlow Elite
1 options
Catalog No. 16-948-961 Supplier Interscience Supplier No. 505007
Add to Cart
Add to Cart
External module with single pump for DilcuFlow
Includes:
  • External module with single pump
  • Dispensing assembly for 6.4 mm dia. glass 5 L bottle (GL45)
Compatible with DiluFlow Elite 1 kg and 5 kg
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) DiluFlow Elite
Sterility Non-sterile