Learn More
EZKEM Pump Tubing Orange/Green 0.015 ID 6x36 12pk
SDP
Catalog No. NC1718712
Encompass_Preferred
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Orange/Green 0.015ID 12/pk
Catalog No. NC1718712 Supplier EZKEM Supplier No. A000345
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Orange/Green 0.015ID 12/pk