Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Reglo ICC Digital Peristaltic Pump, 4-Channel, 12-Roller; 100 to 240 VAC
Catalog No. NC0916780
Encompass
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer¿expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing up to 35 mL/min with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy. Order the three-stop tubing size and formulation to suit your application needs (sold separately). More than 100 options exist ¿ the most popular sizes and formulations can be found under Accessories below
Catalog No. NC0916780 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7800182
Add to Cart
Add to Cart
Request a Quote
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer¿expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensing up to 35 mL/min with the capability of bidirectional flow in each channel. Plus, independent channel calibration minimizes tube-to-tube differences, resulting in high calibration accuracy. Order the three-stop tubing size and formulation to suit your application needs (sold separately). More than 100 options exist ¿ the most popular sizes and formulations can be found under Accessories below