Learn More
Med Vet International "Pump Smooth" IV Set, 105", Anti-Kink Tubing, Baxter Style, J1422B, Each
Catalog No. 5000583672
Encompass_Preferred
High Quality IV sets calibrated to work with VetPro and Heska brand infusion pumps. • Length: 105" • Pump Calibration: Baxter • Injection Y-site
Catalog No. 50-005-83672 Supplier Med Vet International Supplier No. JOR1422B
High Quality IV sets calibrated to work with VetPro and Heska brand infusion pumps. • Length: 105" • Pump Calibration: Baxter • Injection Y-site