Learn More
BURKLE INC Spray bottle with pump vaporizer, tansparent, 8.45 oz. (250 ml)
Supplier:  BURKLE INC 03093025
Encompass_Preferred
The pump spray bottle is a versatile sprayer that atomizes liquids into fine droplets. It saves liquid by distributing it finer, more evenly and over a larger area.
The sprayer is lightweight and easy to use. The actuator is manually pumped at the touch of a button. The pump sprayer requires no propellant and is an environmentally friendly alternative to conventional aerosol spray cans.
The spray bottles are compact and lightweight, ideal for travel. A convenient cap prevents accidental squeezing of the pump and acts as a dust guard.
The clear HDPE bottle provides a good view of the liquid level and easy identification of the contents.
The spray pump is made of durable PP and stainless steel materials and is resistant to a wide range of chemicals.
Each reusable spray bottle is manufactured from robust materials and undergoes rigorous quality control to ensure long life and reliable operation.
Catalog No. 50-000-25985