Learn More
Grainger Red Lion Submersible Deep Well Pump, 0.5 HP, 2 Wire, 115 V, Stainless Steel, 1 ea
Catalog No. 502770853
Encompass_Preferred
This 4 in. submersible deep well pump is designed for the supply of fresh water to rural homes, farms, and cabins that utilize drilled wells with a diameter of 4 in. or greater and depths of up to 250 ft. The pump features a robust stainless steel housing and is engineered for continuous feed operation.
  • Compatible with 4 in. or greater diameter wells
  • Powered by an industry standard 2-wire motor
  • Constructed with a durable stainless steel housing
  • Maximum flow rate of 12 GPM
  • Maximum operating pressure of 80 PSI
  • Includes control box
Catalog No. 50-277-0853 Supplier Grainger Supplier No. 147T19
This 4 in. submersible deep well pump is designed for the supply of fresh water to rural homes, farms, and cabins that utilize drilled wells with a diameter of 4 in. or greater and depths of up to 250 ft. The pump features a robust stainless steel housing and is engineered for continuous feed operation.
  • Compatible with 4 in. or greater diameter wells
  • Powered by an industry standard 2-wire motor
  • Constructed with a durable stainless steel housing
  • Maximum flow rate of 12 GPM
  • Maximum operating pressure of 80 PSI
  • Includes control box