Learn More
Braintree Scientific Pump-to-PC Primary Network Cable, 25 ft.
SDP
Catalog No. 501954512
Encompass
Allows full control over syringe pumps through your computer DB-9 adapter connects to 9-pin serial port (25-pin adapters available upon request) Utilizes the RS-232 communications port of syringe pump Available in 7' and 25' length cable Quick and simple setup **Sold for research purposes only and not for human use**
Catalog No. 50-195-4512 Supplier Braintree Scientific Supplier No. CBLPCPUMP25
Allows full control over syringe pumps through your computer DB-9 adapter connects to 9-pin serial port (25-pin adapters available upon request) Utilizes the RS-232 communications port of syringe pump Available in 7' and 25' length cable Quick and simple setup **Sold for research purposes only and not for human use**