Learn More
World Precision Instrument Pump-to-pump Network Cable, 25 ft
SDP
Catalog No. 502538797
Encompass
The RS-232 Network Cable allows networking of two or more pumps or other devices to a single computer with a quick and simple setup.
Catalog No. 50-253-8797 Supplier World Precision Instrument Supplier No. GNNET25
The RS-232 Network Cable allows networking of two or more pumps or other devices to a single computer with a quick and simple setup.