Learn More
Braintree Scientific Pump-to-Pump Secondary Network Cable
SDP
Catalog No. 501954511
Encompass
Allows networking of two or more syringe pumps to a single computer Quick and simple setup Utilizes the RS-232 communications port of syringe pump Available in 7' and 25' length cable Requires first pump to be connected with primary network cable **Sold for research purposes only and not for human use**
Catalog No. 50-195-4511 Supplier Braintree Scientific Supplier No. CBLNET7
 Restricted Item
Add to Cart
Add to Cart
Allows networking of two or more syringe pumps to a single computer Quick and simple setup Utilizes the RS-232 communications port of syringe pump Available in 7' and 25' length cable Requires first pump to be connected with primary network cable **Sold for research purposes only and not for human use**