Learn More
EZKEM Pump Tubing Orange/Orange0.035 ID 6x36 12pk
SDP
Catalog No. NC0875478
Encompass_Preferred
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Orange/Orange 0.035ID 12/pk
Catalog No. NC0875478 Supplier EZKEM Supplier No. A000349
EZkem’s 2-stop 6″ collar x 36″ length peristaltic pump tubing is used for Ismatec peristaltic pumps in Alpkem, OI Analytical, Latchat and other brands of autoanalyzers for flow injection (FIA) and segmented flow (SFA) analysis. A000352 Orange/Orange 0.035ID 12/pk