Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, Platinum-Cured Silicone, 0.51 mm ID; 50 ft
Catalog No. NC1798602
Encompass
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.
Catalog No. NC1798602 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9559018
Use platinum-cured silicone tubing for high purity applications where outgassing of peroxide product is not allowed.