Learn More
Thermo Scientific Mini Peri-Pump Tubing 3-Stop PVC White / White / White 1.02mm ID (6/pk)
Catalog No. NC0710800 Shop All Thermo Scientific Products
Encompass
Mini Peri-Pump Tubing, 3-Stop PVC 1.02mm (0.040 in.) Tubing White / White / White, Stop Gaps 95mm (3.74 in.), Overall Length 455mm (18 in.), 6 per pack
Catalog No. NC0710800 Supplier Thermo Scientific Supplier No. 842312052401
Mini Peri-Pump Tubing, 3-Stop PVC 1.02mm (0.040 in.) Tubing White / White / White, Stop Gaps 95mm (3.74 in.), Overall Length 455mm (18 in.), 6 per pack