Learn More
Thermo Scientific PUMP TUBING DRAIN ICP 6/PK
Supplier:  Thermo Scientific 842312051521
Encompass
Pump Drain Tube Aqueous; Standard tygon white/white pump tubing.; One pack of six tubes.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC9608337