Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S PharmaPure Pump Tubing, L/S 17, 25 ft
Catalog No. NC0107410
Encompass
PharmaPure Pump Tubing, L/S 17, 25 ft.Ultra low spallation protects fluid purity ¿ ideal for sensitive and costly fluids and for filtration applicationsNontoxic and nonhemolytic with excellent biocompatibilityLong pumping life; pump at continuous pressures up to 40 psiLow extractables and low gas permeabilityMeets USP Class VI criteria
Catalog No. NC0107410 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643517/DEL
PharmaPure Pump Tubing, L/S 17, 25 ft.Ultra low spallation protects fluid purity ¿ ideal for sensitive and costly fluids and for filtration applicationsNontoxic and nonhemolytic with excellent biocompatibilityLong pumping life; pump at continuous pressures up to 40 psiLow extractables and low gas permeabilityMeets USP Class VI criteria