Learn More
Perkin Elmer US LLC Pharmed Three-Stop Peristaltic Pump Tubing - 2.79 mm I.D., Purple/White/Purple, Pkg. 6
Catalog No. 509052081
Encompass_Preferred
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. It can also be used with the AS9X Autosampler and ships as a package of 6.
Catalog No. 50-905-2081 Supplier Perkin Elmer US LLC Supplier No. B3140721
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. It can also be used with the AS9X Autosampler and ships as a package of 6.